Learn More
Bochem™ Ladle Scoop
Bochem™ Ladle Scoop is constructed of Stainless Steel and features a rounded handle.
Brand: Bochem™ 8795
Additional Details : Weight : 0.50000kg
Description
- Stainless Steel
- Rounded handle
- 8mm handle diameter
Specifications
Stainless Steel (18/10) | |
380 mL | |
120 mm |
Rounded Handle | |
400 mm |