Learn More
Invitrogen™ LanthaScreen™ Eu-anti-GFP Antibody, 25 μg
These Europium-labeled anti-GFP antibodies bind specifically to GFP-tagged proteins allowing precise interrogation of protein samples.
Brand: Invitrogen™ A14190
Description
These Europium-labeled anti-GFP antibodies bind specifically to GFP-tagged proteins allowing precise interrogation of protein samples. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
LanthaScreen™ Eu-anti-GFP Antibody Lets You:
- Indirectly label proteins tagged with GFP
- Measure the right kinase
- Interrogate active and non-activated kinases
Eu-anti-GFP Gives You the Benefits of Indirect Labeling
LanthaScreen™ Europium-labeled anti-GFP antibodies can be used for indirect labeling of proteins tagged with GFP. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein negatively impacts protein activity.
Use Eu-anti-GFP Antibody in the Highly Specific LanthaScreen™ Eu Kinase Binding Assay
When LanthaScreen™ Eu-anti-GFP Antibody is used in the LanthaScreen™ Eu Kinase Binding Assay, any signal is the result of a specific interaction of the Eu-anti-GFP antibody with the kinase of interest. Other kinases or phosphatases present without tag or with a different tag produce no signal. Unlike activity assays, the kinase binding assay can be performed with either active or non-active kinase preparations, making it ideal for detection of Type II inhibitors.
Specifications
Kinase Assay | |
Europium | |
RUO | |
Primary | |
Europium labeled anti-GFP antibody in HEPES buffered saline containing 0.1% Triton X-100. Store at -20°C. |
|
Dry Ice | |
LanthaScreen™ |
0.1mg/mL | |
25 μg | |
Kinase Pathway Analysis | |
1 Each | |
Liquid | |
GFP |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.