Learn More
Invitrogen™ LanthaScreen™ Eu-Streptavidin
LanthaScreen™ Eu-Streptavidin
Brand: Invitrogen™ PV6025
This item is not returnable.
View return policy
Description
A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.
Specifications
See the Certificate of Analysis | |
LanthaScreen™ Eu-Streptavidin, 1mg, 0.25mg/mL Concentration, Frozen, 55kDa Molecular Weight, Alexa Fluor™ 647, Europium Dye, Store at –20°C, 340/615, 665nm Excitation/Emission Wavelength, Used For Target & Lead Identification & Validation | |
1 mg | |
Europium-labeled Streptavidin in HEPES buffered saline. Store at -20°C. | |
Dry Ice | |
LanthaScreen™ | |
10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline) | |
Alexa Fluor 647, Europium |
Alexa Fluor 647, Europium | |
55kDa | |
10mM HEPES (pH 7.5), 137mM NaCl and 2.7mM KCl (HEPES buffered saline) | |
340/615, 665 nm | |
Liquid | |
Pharma and biopharma, target and lead identification and validation | |
Streptavidin Conjugate (Fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.