missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody
The LanthaScreen™ Tb-anti-Histone H3K27me3 antibody is a key component of the BacMam Histone H3K27me3 Cellullar Assay (A14159).
74500.00 SEK - 351700.00 SEK
Specifications
Conjugate | Tb (Terbium)/Lanthanide Chelates |
---|---|
Concentration | 3500 nM, 0.5 mg/mL |
Form | Liquid |
Shipping Condition | Dry Ice |
Target Modification State | Methylated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11672049
|
Thermo Scientific™
A14155 |
100 μg |
74500.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11652049
|
Thermo Scientific™
A14152 |
1 mg |
351700.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The LanthaScreen™ Tb-anti-Histone H3K27me3 antibody is a key component of the BacMam Histone H3K27me3 Cellullar Assay (A14159)
- When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 27 trimethylation
- The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 27 of histone H3
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
Tb (Terbium)/Lanthanide Chelates | |
Liquid | |
Methylated |
3500 nM, 0.5 mg/mL | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title