missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-PRAS40 [pSer183] Antibody
For use in combination with the LanthaScreen™ PRAS40 HEK293E cell line (K1528) to evaluate inhibitors in medium to high throughput screening applications.
36660.00 SEK - 210900.00 SEK
Specifications
Conjugate | Tb (Terbium)/Lanthanide Chelates |
---|---|
Excitation/Emission | 340/90, 520 |
Product Line | LanthaScreen™ |
Reactivity | Human |
Readout | End Point |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10525304
|
Thermo Scientific™
PV5578 |
25 μg |
36660.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10093614
|
Thermo Scientific™
PV5579 |
1 mg |
210900.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- PRAS40 (Proline-rich AKT substrate, 40 kDa) is a cytosolic protein that is phosphorylated at position Ser183
- It has been shown to bind to the mTORC1 complex and inhibit downstream signaling (i.e., phosphorylation of 4E-BP1 or p70S6K)
- For use in combination with the LanthaScreen™ PRAS40 HEK293E cell line (K1528) to evaluate inhibitors in medium to high throughput screening applications
Specifications
Tb (Terbium)/Lanthanide Chelates | |
LanthaScreen™ | |
End Point | |
Ser/Thr Kinases |
340/90, 520 | |
Human | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title