missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHA Mouse anti-Human, Mouse, Porcine, Clone: 2E2G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66287-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers.Specifications
LDHA | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00338, P00339, P06151 | |
Ldha | |
LDHA Fusion Protein Ag21417 | |
20 μL | |
Primary | |
Human, Porcine, Mouse | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
2E2G6 | |
Unconjugated | |
LDHA | |
lactate dehydrogenase A, LDH A, LDH M, LDH muscle subunit, LDH1, LDHA, PIG19 | |
Mouse | |
Protein A | |
RUO | |
16828, 3939, 407245 | |
-20°C | |
Liquid |