Learn More
Kartell™ Dispolab™ Push-In Polyethylene Stoppers
Designed to prevent sample spillage caused by jostling and protection if sample is overturned.
Brand:  Kartell™ 0196200
480.96 SEK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 12094677
  • 789.00 SEK / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Description
Designed to prevent sample spillage caused by jostling and protection if sample is overturned.
  • For use with closing Cuvettes with 10 x 10mm square openings
  • Made from LDPE
Specifications
Specifications
Cuvet Cap
LDPE
Closing cuvette with 10 x 10mm square openings
Square
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    22
    Hours
    18
    Minutes
    1
    Seconds
    11
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Kartell™ Dispolab™ Push-In Polyethylene Stoppers > Material: LDPE