missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Acetate Basic, 95+%, Pure, for Sugar Analysis, Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Specifications
Specifications
CAS | 1335-32-6 |
Color | White |
Packaging | Plastic powder jar |
Molecular Formula | C4H10O8Pb3 |
MDL Number | 13041 |
Quantity | 500 g |
Formula Weight | 807.69g/mol |
Physical Form | Powder |
Safety and Handling


Product Identifier
- Lead subacetate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lead(II) Acetate Basic, 95+%, Pure, for Sugar Analysis, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction