Learn More
Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Lead(II) chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 951°C |
Melting Point | 501°C |
Physical Form | Beads |
Quantity | 100 g |
UN Number | UN2291 |
Solubility Information | Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol. |
Formula Weight | 278.1 |
Percent Purity | 99.999% |
Packaging | Ampouled under argon |
Specific Gravity | 5.85 |
Show More |
RUO – Research Use Only