Learn More
Lead Standard Solution, Reagecon™ Product Code.: 12822503
Lead Standard Solution, Reagecon™
BIOPHARMA
Product Code. 12822503 Shop All Reagecon Products
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 12822503
Brand: Reagecon 5001700
Specifications
Packaging 100mL
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5171C24K1 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number5001700
Lot Number-_5171C23M1 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number5001700
Lot Number5171C23A1 Certificate TypeCertificate of Analysis Date29/03/2023 Catalog Number5001700
Lot Number5171C22C1 Certificate TypeCertificate of Analysis Date21/02/2023 Catalog Number5001700
Lot Number5171C21B1 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog Number5001700
Product Content Correction
Product Title
Lead Standard Solution, Reagecon™ >