missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lichenan, ≥95%, For HPLC analysis, MP Biomedicals™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 1402-10-4 |
Color | Gray |
Molecular Formula | C6H10O5 |
Quantity | 1 g |
Synonym | lichenin, 2s,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4,5-triol, licheningehalt |
InChI Key | CXKKSSKKIOZUNR-ZLUOBGJFSA-N |
SMILES | C1=C(C(C(C(O1)CO)O)O)O |
IUPAC Name | (2S,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4,5-triol |
Molecular Weight (g/mol) | 162.141 |
PubChem CID | 439241 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'