missing translation for 'onlineSavingsMsg'
Learn More

Lichenan, MP Biomedicals™

Product Code. 11400485
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Consists of repeating glucose units linked by β-1,3 and β-1,4 glycosidic bonds. Lichenan, MP Biomedicals is a glucose polysaccharide used in the study of biochemical and structural characterization of a β-1,3-1,4-glucanase from Bacillus sub

TRUSTED_SUSTAINABILITY

Specifications

CAS 1402-10-4
Molecular Formula C6H10O5
Quantity 5 g
Synonym lichenin, 2s,3r,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4,5-triol, licheningehalt
Solubility Information Soluble in boiling water and hydrochloride.
InChI Key CXKKSSKKIOZUNR-ZLUOBGJFSA-N
SMILES C1=C(C(C(C(O1)CO)O)O)O
IUPAC Name (2S,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4,5-triol
Molecular Weight (g/mol) 162.141
PubChem CID 439241
Formula Weight 488.44
Physical Form Powder
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'