missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) APC-Cy5.5 Antibody Labeling Kit

Antibody labeling kit

4510.00 SEK - 10570.00 SEK

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate APC/Cy5.5
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy5.5 Antibody Labeling Kit
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18059045
missing translation for 'viewDocuments'
Novus Biologicals™
764-0005
100 μg
4510.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
18769313
missing translation for 'viewDocuments'
Novus Biologicals™
764-0015
1 mg
10570.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
18037565
missing translation for 'viewDocuments'
Novus Biologicals™
764-0030
30 μg
4825.00 SEK
1 set
Please to purchase this item. Need a web account? Register with us today!
18092906
missing translation for 'viewDocuments'
Novus Biologicals™
764-0010
300 μg
7770.00 SEK
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. APC/Cy5.5 is a tandem conjugate. In the Lightning-Link APC/Cy5.5 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5.5. The Cy5.5 emits then the energy received from the APC in the form of long wavelength light at 694nm. The amount of antibody used for labeling should be the same as the pack size of LL-APC-Cy5.5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
APC/Cy5.5
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
APC-Cy5.5 Antibody Labeling Kit
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy5.5 Antibody Labeling Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.