Learn More
Lithium aluminum hydride, 95%, in resealable tin cans, Thermo Scientific Chemicals
306.00 SEK - 1803.00 SEK
Chemical Identifiers
CAS | 16853-85-3 |
---|---|
Molecular Formula | AlH4Li |
Molecular Weight (g/mol) | 37.95 |
MDL Number | MFCD00011075 |
InChI Key | OCZDCIYGECBNKL-UHFFFAOYSA-N |
Synonym | lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
PubChem CID | 21226445 |
IUPAC Name | aluminum;lithium;hydride |
SMILES | [Li+].[AlH4-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10499502
|
Thermo Scientific Acros
332100100 |
10 g | Plastic bag in resealable tin can | N/A | |||||
10459742
|
Thermo Scientific Acros
332101000 |
100 g | Plastic bag in resealable tin can | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
16853-85-3 | |
37.95 | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
21226445 | |
[Li+].[AlH4-] |
AlH4Li | |
MFCD00011075 | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
aluminum;lithium;hydride |
Specifications
125.0°C | |
White to Gray | |
94% min. (LiAlH4) | |
LiAlH4 | |
01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162 | |
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
aluminum;lithium;hydride | |
21226445 | |
95% | |
Lithium Aluminum hydride, In Resealable Tin Cans |
16853-85-3 | |
Crystalline Powder | |
AlH4Li | |
MFCD00011075 | |
15, 344 | |
Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd | |
[Li+].[AlH4-] | |
37.95 | |
37.95 | |
0.9 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-877-9
missing translation for 'rtecsNumber' : BD0100000
missing translation for 'tsca' : TSCA
RUO – Research Use Only