Learn More
Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™, Thermo Scientific Chemicals
1756.42 SEK - 8713.64 SEK
Specifications
Chemical Name or Material | Lithium bis(trimethylsilyl)amide |
---|---|
Name Note | 1.0M Solution in Methyl tert-Butyl Ether |
CAS | 4039-32-1, 1634-04-4 |
Color | Brown to Yellow |
Density | 0.8000g/mL |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10052011
|
Thermo Scientific Acros
345671000 |
100 mL | AcroSeal™ Glass bottle |
1756.42 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10183391
|
Thermo Scientific Acros
345678000 |
800 mL | AcroSeal™ Glass Bottle |
8713.64 SEK
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium bis(trimethylsilyl)amide | |
4039-32-1, 1634-04-4 | |
0.8000g/mL | |
−28°C | |
MFCD00008261 | |
0.8 | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
21 to 25% active base (as LiNSi) |
1.0M Solution in Methyl tert-Butyl Ether | |
Brown to Yellow | |
55.0°C to 56.0°C | |
C6H18LiNSi2 | |
04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.33 | |
167.33 | |
Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-725-6
missing translation for 'tsca' : TSCA
- Lithium bis(trimethylsilyl)amide, 1.0M solution in methyl tert-butyl ether
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Methyl tert-butyl ether, Lithium bis(trimethylsilyl)amide