missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Litmus, Indicator, pH 5-8, for titration, Honeywell Fluka™
Indicator, pH 5-8, for titration
Brand: Honeywell-Fluka L7382-100G
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Litmus | |
1393-92-6, 64-17-5 | |
≥96.0000% | |
I2 | |
Analytical Reagent | |
100 g |
Indicator, pH 5-8, for titration | |
≤4.0000%, ≤100.0000% | |
I2 | |
MFCD00011355 | |
Glass Bottle |