Learn More
Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO4
For optimized expression of protein in CHO cells
1465.00 SEK - 2410.00 SEK
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | With Additives | Without Additives | Price | Quantity & Availability | |||
11675140
|
Lonza™
04-919Q |
1 L | None | No Phenol Red, No 0.1% Pluronic F-68, No Glutamine, No Hypoxanthine, No Thymidine |
2410.00 SEK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11645160
|
Lonza™
BE12-029Q |
1 L | Phenol Red, 0.1% Pluronic F-68 | No Hypoxanthine, No Thymidine, No Glutamine | |||||
Description
- ProCHO-4 is designed for the concurrent transition of adherent CHO cells to serum-free and suspension culture; supports faster doubling times
Specifications
2° to 8°C | |
CHO | |
Liquid | |
1 L |