missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Ham's F-12 Medium
Designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. BioWhittaker™ Ham's F-12 Medium has been used for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells and rat, chicken and rabbit embryos.
Brand: Lonza™ BE12-615F
This item is not returnable.
View return policy
Description
- Nutrient mixture designed to cultivate a wide variety of mammalian and hybridoma cells when used with serum in combination with hormones and transferrin
- F-12 medium is based on Ham’s F-10 medium with increased concentrations of choline, inositol, putrescine and many amino acids
- Manufactured in accordance with cGMP regulations
- Formulations used for standard liquid media products are those recommended by the Tissue Culture Association
Specifications
2° to 8°C | |
Sterile | |
7.07 to 7.4 |
Ham's F-12 | |
Sterile-filtered through two consecutive 0.2 μm filters | |
500 mL |