Learn More
Lonza™ BioWhittaker™ ProFreeze™ NAO Freezing Medium
Brand:  Lonza™ 12-769E
Product Code. 11695250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
ProFreeze™-CDM, NAO, Chemically Defined Freeze Medium (2X) is universally suitable for cryopreserving many cell types in the absence of fetal bovine serum (FBS). However, it is used to greatest advantage with cells cultured in a serum-free and animal component-free environment. This protein-free freezing medium contains no animal derived components, insulin, or hydrolysate, and maintains high cell viability upon recovery from frozen storage.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Liquid
100 mL
2 X
Freezing Medium