Learn More
Lonza™ DNA Standards for Pulsed Field Gel Electrophoresis
For size estimation of megabase DNA separated by pulsed field gel electrophoresis (PFGE)
Brand:  Lonza™ 50401
 View more versions of this product
Product Code. 11520797
  • 6060.00 SEK / Pack of 5
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
  • Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
  • SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
  • Store at 2° to 8°C for up to 12 months
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA Molecular Weight Marker/Ladder
Size estimation of megabase DNA separated by pulsed field Gel electrophoresis
5 Plugs
lambda ladder, 50mM EDTA, 10mM [Tris(hydroxymethyl)aminoethane], pH 8, 5μg/mL Proteinase K 0.01% N-lauroyl Sarcosine
18
2° to 8°C