Learn More
Lovibond™ Primary Chlorine Standard Kit for Colorimeters, Cole-Parmer™
Validate the calibration and accuracy of your instrument
Brand:  Cole Parmer 99560-90
Additional Details : Weight : 0.50000kg
Product Code. 15212056
  • 645.00 SEK / Each
Estimated Shipment: 11-09-2024
to see stock.
Description
Description
  • Complies with new regulations specified in US EPA Method 334.0.
  • Ideal for laboratory and on-line chlorine measurements.
  • Chlorine standard ampoule, 97mL of zero chlorine water and ampoule breaker.
  • Everything you need for easy preparation of a 1.5mg/L NIST traceable chlorine standard.
  • Simple, time saving and practical kit to help comply with the new regulations
Specifications
Specifications
1Box/Kit
Chlorine standard ampoule, 97mL of zero chlorine water, and ampoule breaker
Colorimeter