Learn More
LRPAP Antibody (7F1), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Bio-Techne NBP3-12047AF700
Product Code. 18324033
  • 5815.00 SEK / 0.10mL
Explore more special offers
Description
Description
LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
LRPAP
Monoclonal
Alexa Fluor 700
50mM Sodium Borate
Rabbit
Protein A purified
RUO
4043
Store at 4C in the dark.
IgG κ
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
7F1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP
This antibody was raised by immunising mice with human placental LRPAP.
0.1 mL
Primary
Human
Purified