missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC63 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558918
This item is not returnable.
View return policy
Description
Immunogen sequence: TATLTLRVTE FPGFVSLPTP VLPRKPHRQS VIETLVTENG NIESVPKQIP PRPPEGLTKT EKIESEIHVV RGEGFKTVAA TRYETITAMT NLAIVNCQVY GRN Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 47%.
LRRC63 is a protein coding gene.Specifications
LRRC63 | |
Polyclonal | |
Unconjugated | |
LRRC63 | |
leucine rich repeat containing 63; leucine-rich repeat-containing protein 63; LRRC63 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q05C16 | |
LRRC63 | |
Recombinant protein corresponding to Human LRRC63 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only