missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTB4R2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533912
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%) Mouse, Rat, Bat (88%) Elephant (81%).
BLTR2 has been reported in humans in peripheral blood leukocytes, mononuclear lymphocytes, and spleen. ESTs have been isolated from a broad array of human libraries, including brain, eye, fetal lung/testis/B-cell, heart, kidney, melanocyte/uterus/fetal heart, testis, tonsil, and uterus libraries.Specifications
LTB4R2 | |
Polyclonal | |
Unconjugated | |
LTB4R2 | |
5830462O07Rik; BLT2; Blt-2; BLT2R; BLTR2; Hblt2; JULF2; KPG_004; Leukotriene B4 receptor 2; leukotriene B4 receptor BLT2; LTB4 receptor JULF2; LTB4-R 2; LTB4R2; LTB4-R2; LTB4-R-2; NOP9; Seven transmembrane receptor BLTR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56413 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NPC1 | |
LTB4R2 | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human LTB4R2. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |