missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTB4R2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550824
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total BLT2 protein.
BLTR2 has been reported in humans in peripheral blood leukocytes, mononuclear lymphocytes, and spleen. ESTs have been isolated from a broad array of human libraries, including brain, eye, fetal lung/testis/B-cell, heart, kidney, melanocyte/uterus/fetal heart, testis, tonsil, and uterus libraries.Specifications
LTB4R2 | |
Polyclonal | |
Unconjugated | |
LTB4R2 | |
5830462O07Rik; BLT2; Blt-2; BLT2R; BLTR2; Hblt2; JULF2; KPG_004; Leukotriene B4 receptor 2; leukotriene B4 receptor BLT2; LTB4 receptor JULF2; LTB4-R 2; LTB4R2; LTB4-R2; LTB4-R-2; NOP9; Seven transmembrane receptor BLTR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56413, 57260 | |
-20°C | |
Liquid |
Western Blot | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9JJL9, Q9NPC1 | |
LTB4R2 | |
Synthetic peptide corresponding to residues near the C terminal of human leukotriene B4 receptor 2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |