Learn More
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67293-1-IG-20UL
This item is not returnable.
View return policy
Description
The luciferase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of luciferase. This antibody recognizes gaussia princeps antigen. The luciferase antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
luciferase | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
IgG2a |
Western Blot | |
2D2E1 | |
Unconjugated | |
20 μL | |
Plant |