Learn More
Lumicitabine, MedChemExpress
MedChemExpress Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.
Brand: Medchem Express HY-12983A
This item is not returnable.
View return policy
Description
Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.Specifications
Lumicitabine | |
White | |
C18H25ClFN3O6 | |
ALS-008176 ALS-8176 | |
ClC[C@@]1(COC(C(C)C)=O)[C@@H](OC(C(C)C)=O)[C@@H](F)[C@H](N2C(N=C(N)C=C2)=O)O1 | |
433.86 | |
Research | |
COVID-19-anti-virus |
1445385-02-3 | |
Research | |
5 mg | |
DMSO : ≥ 50 mg/mL (115.24 mM) | |
433.86 | |
98.02% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.