Learn More
Lumicitabine, MedChemExpress
MedChemExpress Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.
Brand:  Medchem Express HY-12983A
Encompass_Preferred
 View more versions of this product
Product Code. 16491319
  • 2637.43 SEK / 1mg
Explore more special offers
Description
Description
Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.
Specifications
Specifications
Lumicitabine
White
C18H25ClFN3O6
ALS-008176 ALS-8176
ClC[C@@]1(COC(C(C)C)=O)[C@@H](OC(C(C)C)=O)[C@@H](F)[C@H](N2C(N=C(N)C=C2)=O)O1
433.86
Research
COVID-19-anti-virus
1445385-02-3
Research
1 mg
DMSO : ≥ 50 mg/mL (115.24 mM)
433.86
98.02%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.