missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lupeol

Product Code. 16637032
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545-47-1
Molecular Formula C30H50O
Molecular Weight (g/mol) 426.73
MDL Number MFCD00017351
InChI Key MQYXUWHLBZFQQO-JGGBMTAGSA-N
IUPAC Name (1R,3aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a,5a,5b,8,8,11a-hexamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
SMILES CC(=C)[C@@H]1CC[C@]2(C)CCC3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12

Specifications

Alpha Vector LUPEOL
Melting Point 206°C to 210°C
Color White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
Solubility Information practically insoluble aq. soln., freely soluble in ether, benzene, petroleum ether, warm alcohol, practically insoluble in chloroform, methanol
Formula Weight 426.72
Physical Form Powder
Chemical Name or Material Lupeol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Lupeol >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.