missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY 288513, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective CCK2 receptor antagonist (IC50 values are 16 and > 30,000 nM for CCK2 and CCK1 respectively). Displays anxiolytic and antipsychotic properties in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 436.3 |
Percent Purity | >98% |
Chemical Name or Material | LY 288513 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'