Learn More
Macherey-Nagel™ Lysis Buffer T1
Lysis buffer for isolation and clean-up of genomic DNA samples using compatible kits (e.g., NucleoSpin™ Tissue kits, NucleoMag™ DNA Food).
Brand:  Macherey-Nagel™ 740940.100
 View more versions of this product
Product Code. 15899137
  • 710.00 SEK / 100mL
Explore more special offers
This item is not returnable. View return policy
Specifications
Colorless
Compatible kits (e.g. NucleoSpin Tissue kits, NucleoMag DNA Food)
7.5 to 8.5
Odorless
Liquid
Lysis Buffer T1
Up to 2%
Chemicals mixture in water
100 mL
Room Temperature (18°C to 25°C)
Isolation and cleanup of DNA