missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Rabbit anti-Human, Clone: 9F0W6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16466-100UL
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western BlotSpecifications
Lysyl Oxidase Homolog 2/LOXL2 | |
Monoclonal | |
Unconjugated | |
EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
A synthetic peptide corresponding to a sequence within amino acids 675-774 of human Lysyl Oxidase Homolog 2/LOXL2 (Q9Y4K0). | |
100 μg | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
4017 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
9F0W6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction