missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl acetate, 97%, Thermo Scientific Chemicals

Product Code. 11303616
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 122-46-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00041910
InChI Key OTGAHJPFNKQGAE-UHFFFAOYSA-N
Synonym m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
PubChem CID 67406
IUPAC Name (3-methylphenyl) acetate
SMILES CC(=O)OC1=CC=CC(C)=C1

Specifica

Density 1.05
Boiling Point 100°C (14 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.502
Quantity 100 g
Beilstein 1862796
Merck Index 14,2583
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material m-Tolyl acetate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'