Learn More
Applied Biosystems™ M13 Human, Reverse Primer, Desalted
M13 Human, Reverse Primer, Desalted
Brand: Applied Biosystems™ A15634
This item is not returnable.
View return policy
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Specifications
M13 Human Reverse Desalted Primer, 25 nmole, Solid, For PCR and Sanger Sequencing | |
Desalted | |
25 nmol | |
Solid |
M13 Human Reverse DS Primer | |
Reverse | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.