Learn More
MP Biomedicals™ MacConkey Agar
MacConkey Agar is powder media used for isolating enteric bacilli.
Brand:  MP Biomedicals™ 091006017
Product Code. 11470215
  • 1165.00 SEK / 500g
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Medium used for the isolation of enteric bacilli based on the fermentation of lactose.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Powder
MacConkey Agar
500 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.