Learn More
MP Biomedicals™ MacConkey Agar
MacConkey Agar is powder media used for isolating enteric bacilli.
Brand: MP Biomedicals™ 091006017
This item is not returnable.
View return policy
Description
- Medium used for the isolation of enteric bacilli based on the fermentation of lactose.
Specifications
Powder | |
MacConkey Agar | |
500 g |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.