missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ MACROMAN™ Pipette Adaptor
This pipette adaptor is a silicone replacement part for the MACROMAN™ pipette controller.
Brand: Gilson™ F107211
This item is not returnable.
View return policy
Description
- For use with MACROMAN™ pipette controllers
- Can be autoclaved at 121°C for 20 minutes at 1 bar relative
- Supplied individually
Specifications
Autoclavable (121°C) | |
Adapter | |
1 |
Silicone | |
MACROMAN™ Pipette Controller |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction