Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
Brand: Cytiva MAP003C38
14763.00 SEK valid until 2025-12-31
Use promo code "24320" to get your promotional price.
This item is not returnable.
View return policy
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
Gray | |
PES | |
Non-sterile | |
5 to 20 mL (Sample) |
EFA: 7.2 cm2 | |
3 kDa | |
100/Pk. |
Product Content Correction
Product Title
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane > 3 kDa; Gray; 100/Pk.
Spot an opportunity for improvement?Share a Content Correction