missing translation for 'onlineSavingsMsg'
Learn More

MAD1 Monoclonal Antibody (9B10), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA15830

Product Code. 11514373

  • 4055.00 SEK / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAD1
Monoclonal
0.9 mg/mL
PBS with 0.1% sodium azide
Q9Y6D9
MAD1L1
Recombinant full length Mad1 protein.
50 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunocytochemistry, Immunoprecipitation
9B10
Unconjugated
MAD1L1
AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W
Mouse
Protein A
RUO
8379
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers