missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide, 98%, pure, anhydrous, Thermo Scientific Chemicals

262.33 SEK - 344.44 SEK

Chemical Identifiers

CAS 7789-48-2
Molecular Formula Br2Mg
Molecular Weight (g/mol) 184.11
MDL Number MFCD27995637
InChI Key OTCKOJUMXQWKQG-UHFFFAOYSA-L
Synonym magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent
PubChem CID 82241
IUPAC Name magnesium;dibromide
SMILES [Mg++].[Br-].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10351290
View Documents
Thermo Scientific Acros
413390010
1 g Glass bottle
262.33 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10024157
View Documents
Thermo Scientific Acros
413390050
5 g Glass bottle
344.44 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-48-2
184.11
OTCKOJUMXQWKQG-UHFFFAOYSA-L
82241
[Mg++].[Br-].[Br-]
Br2Mg
MFCD27995637
magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent
magnesium;dibromide

Specifications

711.0°C
97.5 to 102.5% (Argentometry)
MgBr2
02,254; 06,270; 08,273; 10,14; 15,194; 16,199
magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent
OTCKOJUMXQWKQG-UHFFFAOYSA-L
magnesium;dibromide
82241
98%
Magnesium bromide, Anhydrous
7789-48-2
Br2Mg
MFCD27995637
15,5723
Solubility in water: 1015g/L (20°C). Other solubilities: slightly soluble in alcohol
[Mg++].[Br-].[Br-]
184.11
184.11
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 232-170-9

Product Identifier
  • Magnesium bromide, anhydrous

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.