missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium ethoxide, Mg 21-22%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Magnesium ethoxide is used as olefin polymerization catalyst carrier for polypropylene, polyethylene and precision ceramic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 270°C (decomposition) |
Color | White |
Density | 1.01 g/mL |
Odor | Odorless |
Linear Formula | Mg(OCH2CH3)2 |
Quantity | 1 kg |
UN Number | UN3205 |
Beilstein | 3678841 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'