missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
384.00 SEK - 1119.84 SEK
Chemical Identifiers
CAS | 7782-75-4 |
---|---|
Molecular Formula | H7MgO7P |
Molecular Weight (g/mol) | 174.33 |
MDL Number | MFCD00150390 |
InChI Key | OKIWLDVQGKRUNR-UHFFFAOYSA-L |
Synonym | magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg |
PubChem CID | 16204530 |
IUPAC Name | magnesium;hydrogen phosphate;trihydrate |
SMILES | O.O.O.[Mg++].OP([O-])([O-])=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-75-4 | |
174.33 | |
OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
16204530 | |
O.O.O.[Mg++].OP([O-])([O-])=O |
H7MgO7P | |
MFCD00150390 | |
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg | |
magnesium;hydrogen phosphate;trihydrate |
Specifications
7782-75-4 | |
H7MgO7P | |
MFCD00150390 | |
(0.1 M in 5 M HCl) Clear colorless | |
O.O.O.[Mg++].OP([O-])([O-])=O | |
174.33 | |
174.34 | |
Biochemical | |
2.1300g/mL |
97.5% min. (Complexometry) | |
MgHPO4·3H2O | |
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg | |
OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
magnesium;hydrogen phosphate;trihydrate | |
16204530 | |
98% | |
2.13 | |
Magnesium hydrogen phosphate trihydrate |
Safety and Handling
Product Identifier
- Magnesium hydrogen phosphate trihydrate
RUO – Research Use Only