Learn More
Mettler Toledo™ Magnetic Stirrer EasyMix
Magnetic stirrer EasyMix for use with SevenDirect meters. The EasyMix can be controlled manually or automatically by the SevenDirect meter.
Brand: Mettler Toledo™ 30671570
3574.78 SEK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- The Right Stirrer
- A dedicated stirrer for the SevenDirect generates key advantages: programmable stirring and a perfect fit with the EasyPlace™ electrode arm
- Program Consistent Conditions
- Program the stirring speed from the pH meter to ensure consistent conditions for all operators
- Define preconditioning and waiting periods
- Perfect Fit Saves Space
- The EasyMix magnetic stirrer is a perfect fit with the EasyPlace electrode arm
- Together, they form a tidy and compact setup
- Reliably Stir Large Volumes
- The larger the sample, the higher the risk of inhomogeneity
- Rely on the strength of the EasyMix to stir efficiently up to 10 L of sample volume
Specifications
Magnetic Stirrer, EasyMix | |
EasyMix |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%