Learn More
3M™ Maintenance-Free Reusable Half Mask 4000+ Series
Available to help prolong the life of the main particulate filter for the 3M™ Maintenance-Free Reusable Mask 4000+ Series. Traps larger spray particles before they reach and clog the filters.
Brand: 3M™ 7100113104
5820.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.50000kg
Description
- 3M™ Overspray Guard 400+ prolongs respirator service life by reducing clogging of the particulate filters during paint spray applications
- Keeps filters clean during paint spray applications
- Traps larger spray particles before they reach and clog the filters
- For use with 3M™ Reusable Respirator 4000+ Series
Specifications
Reusable | |
3M™ Maintenance-Free Reusable Mask 4000+ Series |