missing translation for 'onlineSavingsMsg'
Learn More
Maleic Acid, Technical, MP Biomedicals Product Code.: 11309099

Maleic Acid, Technical, MP Biomedicals

Product Code. 11309099 Shop All MP Biomedical Products
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11309099

Brand: MP Biomedicals 0210223201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 110-16-7
Molecular Formula C4H4O4
MDL Number MFCD00063177
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES OC(=O)\C=C\C(O)=O
IUPAC Name (2E)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
PubChem CID 444266
ChEBI CHEBI:18300
Formula Weight 116.07
Grade Technical
Quantity 1 kg
Chemical Name or Material Maleic Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Maleic Acid, Technical, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.