Learn More
Macherey-Nagel™ Manual Ergonomic Decapper
3020.00 SEK
Specifications
No. per Pack 1 Each
Type Manual Ergonomic Decapper
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
12898585
View Documents
Macherey-Nagel™
735311
For 11mm Aluminum Crimp Caps
3020.00 SEK
Each
12808595
View Documents
Macherey-Nagel™
735320
For 20mm Aluminum Crimp Caps
3020.00 SEK
Each
Specifications
1 Each
Manual Ergonomic Decapper
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Manual Ergonomic Decapper