missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCO Rabbit anti-Human, Clone: PLK1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-12048-0.2MG
This item is not returnable.
View return policy
Description
MARCO Monoclonal antibody specifically detects MARCO in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
MARCO | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
The antibody was raised by immunising BALB/c mice with human alveolar macrophages (AMs) and subsequent creation of hybridomas. | |
0.2 mg | |
Primary | |
Human | |
IgG κ |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
PLK1 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
8685 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction