Learn More
Masterflex™ Translucent C-Flex Braided Tubing
Brand:  Masterflex™ 06433-07
 View more versions of this product
Product Code. 15262700
  • 3440.00 SEK / 25ft
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Specifications
FDA compliant, and complies with USP Class VI standards
12.8 mm
19.05 mm
For use with cell cultures, vessel and tank transfer, pump applications and cold storage.
1 Pack
135°C
Autoclavable
C-Flex
7.62 mm
-45°C
Translucent