Learn More
MBL Volumetrics™ Burette Tubes
Replacement part for modular Burette tube
Brand: MBL BRB276P
243.20 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.23000kg
Specifications
50 mL | |
Glass | |
Each | |
Buret Tube |