Learn More
MCP-5 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
2205.00 SEK - 19580.00 SEK
Specifications
Antigen | MCP-5 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17843583
|
Invitrogen
500-P61-50UG |
50 μg |
2205.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17813023
|
Invitrogen
500-P61-100UG |
100 μg |
2830.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17823353
|
Invitrogen
500-P61-1MG |
1 mg |
19580.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-5 (CCL12). Anti-Murine MCP-5 (CCL12)-specific antibody was purified by affinity chromatography employing an immobilized Murine MCP-5 (CCL12) matrix. Sandwich ELISA: To detect Murine MCP-5 (CCL12) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine MCP-5 (CCL12) (500-P61Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MCP-5 (CCL12). Western Blot: To detect Murine MCP-5 (CCL12) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine MCP-5 (CCL12) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.Specifications
MCP-5 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Q62401 | |
20293 | |
E.coli-derived Recombinant Murine MCP-5 (CCL12) | |
Antigen affinity chromatography | |
Ccl12 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
C-C motif chemokine 12; Ccl12; chemokine (C-C motif) ligand 12; MCP-1-related chemokine; Mcp5; MCP-5; Monocyte chemoattractant protein 5; monocyte chemotactic protein 5; Scya12; small inducible cytokine A12; small-inducible cytokine A12 | |
Ccl12 | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.