Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
Brand: Hirschmann™ 2950170
This item is not returnable.
View return policy
Description
- Class A
- Pursuant to the rules of the pre-packing regulation
- Certified conformity
- Blue graduation
- Non-ground, rimmed
- Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
- Calibrated for IN
Specifications
25 mL | |
0.5 mL | |
Yes |
23 to 27 mL | |
110 mm | |
A |